• USA Home
  • Z153001 - Thermolyne® 45500 temperature controller
EMAIL THIS PAGE TO A FRIEND
Z153001 Aldrich
Thermolyne® 45500 temperature controller
AC/DC input 240 V AC
Purchase
Properties
Related Categories Heating, Heating Controllers, Labware More...
mfr. no.   Barnstead International CN45510
Description
General description
Excellent for use with heating mantles and tapes, small water baths, and small heating elements. Utilizes time-on/time-off stepless input control. Built-in cycle and power-on light. One standard, single-duplex outlet at rear of housing. Compensates for ambient temperature and line voltage fluctuations. Complete with 5 ft, 3-wire cord and plug. Max. load current: 15A.
Legal Information
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?