• USA Home
  • Z153206 - Cochranes molecular models
EMAIL THIS PAGE TO A FRIEND
Z153206 Aldrich
Cochranes molecular models
micrit, single kit with booklet
Synonym: chemistry model sets, molecular model sets
Purchase
Properties
Related Categories Cochranes models, Labware, Molecular Models, Molecular Models and Educational Aids
feature   no. of atoms> 1,700
  no. of bonds> 1,000
mfr. no.   Cochranes, 0109
scale   2 cm = 0.1 nanometer (1 Å), Minit
  3 cm = 0.1 nanometer (1 Å), Orbit
Description
General description
An extensive model system offering 15 atomic bonding configurations in 12 colors of plastic atoms. The carbon atom is represented in nine configurations: linear, trigonal (in five angle variations), tetrahedral, octahedral, trigonal-bipyramidal. Models come in two sizes: Orbit and Minit. Spare atom centers and bonding tubes are available for all model kits.
Please see Technical Information Bulletin AL-183 for a listing of these items, available on our website at sigma-aldrich.com
For building macro pharmacological models. Space-filling spheres and molded bond tubes at a scale of 1.25 cm = 100 pm.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?