EMAIL THIS PAGE TO A FRIEND
Z153850 Aldrich
Mininert® Valve
Joint valve, Joint: ST/NS 19/22
Purchase
Description
Application
Mininert valves are leak-tight closures for vials and other containers. They are available in both crimp and screw thread versions. Only PTFE and glass are in contact with the contents of the container.
They are the ideal closure for the following applications:
• Calibration standards
• Air- or moisture-sensitive fluids
• Derivatizating reagents
• Volatile chemicals
General description
These valves are very easy to use:
      1. Push the green button to open the valve
      2. Insert the needle through the septum and take sample
      3. Remove the needle and push to close
Benefits include:
• PTFE construction for inertness. PTFE is useful when working when acids and organic solvents
• Good sealing ability for containing most volalite liquids and gases at low pressures
• Mininert valves can be used at temperatures up to 105°F
• 0.036 in. (0.9 mm) opening accepts up to 20-gauge needles
• Silicone septa
Legal Information
Mininert is a registered trademark of Valco Instruments Co. Inc.
Price and Availability
All labs need water
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?