• USA Home
  • Z154385 - Ace Trubore® water-cooled stirrer bearing
EMAIL THIS PAGE TO A FRIEND
Z154385 Aldrich
Ace Trubore® water-cooled stirrer bearing
Joint: ST/NS 55/50
Purchase
Properties
Related Categories Ace, Ace Stirrer Bearings, Ace Stirrers, Glass Bearing Adapter, Glass Stirring Equipment,
Joint   Joint: ST/NS55/50
feature   fits shaft o.d. 19 mm
mfr. no.   Ace Glass 805909
Description
General description
Top recommended stirring speed of 1,500 rpm when lubricated with Stir-Lube, Z12,603-9.
Legal Information
Trubore is a registered trademark of Ace Glass, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?