• USA Home
  • Z154423 - Ace Trubore® PTFE 4-piece stirrer bearing
EMAIL THIS PAGE TO A FRIEND
Z154423 Aldrich
Ace Trubore® PTFE 4-piece stirrer bearing
Joint: ST/NS 35/25, fits shaft o.d., 10 mm
Purchase
Properties
Related Categories Ace, Ace Stirrer Bearings, Ace Stirrers, Glass Bearing Adapter, Glass Stirring Equipment,
Joint   Joint: ST/NS35/25
feature   fits shaft o.d. 10 mm
mfr. no.   Ace Glass 806660
Description
General description
Self-aligning, lubricant-free PTFE Trubore bearing for use with SS or polished glass stirring shafts at approximately 200rpm. Consists of PTFE inner Trubore bearing, FETFE® compression saddle O-ring, glass-reinforced bushing and lock-nut with FETFE O-ring, and threaded glass adapter. The lock permits stirring in either direction without fear of unthreading. The compression saddle O-ring maintains constant force with little attention. Vacuum to 1 mm Hg. Not recommended for use with ground-glass shafts.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Trubore is a registered trademark of Ace Glass, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?