EMAIL THIS PAGE TO A FRIEND
Z154571 Aldrich
Positioning collar
PTFE/Glass composite, fits shaft o.d., 19 mm
Purchase
Properties
Related Categories Ace, Ace Stirrer Shafts, Ace Stirrers, Glass Stirrer Shafts, Glass Stirring Equipment,
material   PTFE gasket
feature   fits shaft o.d. 19 mm
mfr. no.   Ace Glass 812720
Description
General description
Designed for use with stirring shafts to position shaft in bearing and prevent it from dropping into flask. Gasket prevents scratching bearing and acts as a dust cover.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?