• USA Home
  • Z154806 - VARITEMP® heat gun
EMAIL THIS PAGE TO A FRIEND
Z154806 Aldrich
VARITEMP® heat gun
AC input 220-240 V AC
Purchase
Properties
Related Categories Heat Guns, Heating, Labware More...
feature   CE compliant: no
mfr. no.   , VT-752C
availability   not available in EU
parameter   ≤750 °F temperature (400 °C)
output   7 A
Description
General description
Dial-in and lock temperature control (ambient to maximum); 3-position rocker switch (off/cold/hot); rubber-backed base with 90° rotation; heavy-duty 3-wire line cord and plug on 120 V unit (220/240 V unit supplied without plug); mica-insulated ceramic heating element; double-jacketed die-cast aluminum housing.
Legal Information
VARITEMP is a registered trademark of Master Appliance Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?