• USA Home
  • Z155136 - Protectoseal® safety disposal can
EMAIL THIS PAGE TO A FRIEND
Z155136 Aldrich
Protectoseal® safety disposal can
capacity 5 gal, 316 stainless steel
Purchase
Properties
Related Categories Disposal Containers, Flammable and Corrosive Material Disposal Contaners, Labware, Metal Safety Containers, Safety Containers,
material   316 stainless steel
mfr. no.   Protectoseal
availability   not available in EU
H × O.D.   20 in. × 12 in.
capacity   5 gal
Description
General description
Use for safe, convenient disposal of flammable or combustible liquids.
• Stainless steel or terneplated steel
• Wide (35/8 in.) opening permits use without funnel; spout cap opens to 80°
• Perforated SS metal flash arrester prevents ignition of contents
• Spring-loaded, self-closing, leakproof cap minimizes evaporation and provides automatic pressure relief
• FM approved
Legal Information
Protectoseal is a registered trademark of Protectoseal Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?