• USA Home
  • Z155403 - Stainless steel pan Type 304
EMAIL THIS PAGE TO A FRIEND
Z155403 Aldrich
Stainless steel pan Type 304
standard weight, L × W × H 6.9 in. × 6.4 in. × 6 in.
Purchase
Properties
Related Categories Containers, Labware, Pans and Trays More...
material   stainless steel container
mfr. no.   Polarware, E06066
L × W × H   6.9 in. × 6.4 in. × 6 in.
capacity   2.625 qt
Description
General description
Wide assortment of standard and heavy duty, seamless trays have coved corners for easy sterilization. Covers have recessed strap handles for ease in handling and storing.
Supplied without cover.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
accessory
Product #
Description
Add to Cart
Z155411 Solid cover for type 304 SS pan, Fits pans 6.9 in. x 6.4 in.
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?