• USA Home
  • Z156884 - Justrite® round Type 1 can
EMAIL THIS PAGE TO A FRIEND
Z156884 Aldrich
Justrite® round Type 1 can
capacity 5 gal
Purchase
Properties
Related Categories Justrite, Justrite Safety, Labware, Manufacturer Browser, Plastic Safety Containers,
material   cadmium-plated fitting
  high-density polyethylene round
mfr. no.   Justrite 14501
H × O.D.   16 in. × 12 3/4 in.
capacity   5 gal
Description
General description
Tough, seamless, resilient red HDPE body safety cans resist physical abuse and are impervious to most solvents, fuels, and corrosive chemicals. Self-closing pressure-relief cap and SS flame arrester. Withstands 50 psig without rupture or structural damage. FM approved.
Legal Information
Justrite is a registered trademark of Justrite Manufacturing Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
Z156914 Justrite® nonmetallic wide-spout disposal can, capacity 5 gal
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?