• USA Home
  • Z156906 - Justrite® nonmetallic wide-spout disposal can
EMAIL THIS PAGE TO A FRIEND
Z156906 Aldrich
Justrite® nonmetallic wide-spout disposal can
capacity 2 gal
Purchase
Properties
Related Categories Disposal Containers, Flammable and Corrosive Material Disposal Contaners, Justrite, Justrite Safety, Labware,
mfr. no.   Justrite 14722
H × O.D.   14 3/4 in. × 12 in.
capacity   2 gal
Description
General description
Handles hazardous spent liquids. Resilient red HDPE resists physical abuse and is impervious to most solvents, fuels, and corrosive chemicals. Self-closing cap, 35/8 in. pouring spout, pressure relief valve, and SS flame arrester. Withstands 50 psig without rupture or structural damage. FM approved. Cadmium-plated fittings.
Legal Information
Justrite is a registered trademark of Justrite Manufacturing Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?