• USA Home
  • Z158178 - Ace Buchner funnel with hose connection and 24/40 joint
EMAIL THIS PAGE TO A FRIEND
Z158178 Aldrich
Ace Buchner funnel with hose connection and 24/40 joint
capacity 140 mL, porosity 4-8 μm
Purchase
Properties
Related Categories Ace, Ace Buchner Funnel with Hose Connection and 24/40 Joint, Ace Funnels, Buchner Funnels, Fritted, Funnels,
mfr. no.   Ace Glass 718420
capacity   140 mL
disc diam.   65 mm
pore size   4-8 μm porosity
Description
General description
With glass frit.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
Z546976 Aldrich® Buchner funnel with joint, capacity 140 mL, disc diam. 65 mm, porosity 4-8 μm, Joint: ST/NS 24/40
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?