• USA Home
  • Z158453 - Scienceware® bench lab rack
EMAIL THIS PAGE TO A FRIEND
Z158453 Aldrich
Scienceware® bench lab rack
L × W × H 18 in. × 7 in. × 12 1/2 in.
Purchase
Properties
Related Categories Bel-Art, Bel-Art Cleaners, Cleaners, Dryers and Racks, Labware,
mfr. no.   Bel-Art
L × W × H   18 in. × 7 in. × 12 1/2 in.
Description
General description
Molded PE. One-piece base has trays with scalloped edges for holding pipettes, glass rods and tubes. Sloping back holds 22 white-nylon pegs mounted at 45° angle.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z673463 LAB-AIRE® II non-electrical drying racks, benchtop or wall mounted, single sided, 1 tier
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?