• USA Home
  • Z158526 - Sellstrom® 20/20 safety glasses
EMAIL THIS PAGE TO A FRIEND
Z158526 Aldrich
Sellstrom® 20/20 safety glasses
clear polycarbonate lens
Purchase
Properties
Related Categories Eye Protection Accessories, Eye Protection Equipment, Labware, Safety Equipment
material   clear polycarbonate lens
size   64 mm
Description
General description
Scratch resistant. Form-fit protects front, top, and sides of face. Excellent peripheral vision. Meets ANSI Z87.1-1989 requirements.
No UV protection, no IR protection
Legal Information
Sellstrom is a registered trademark of Sellstrom Manufacturing Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
Z288101 Vision Series 3000 racheting safety glasses, clear Lens, yellow temple arm
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?