• USA Home
  • Z158801 - Walters vacuum manifold
EMAIL THIS PAGE TO A FRIEND
Z158801 Aldrich
Walters vacuum manifold
Purchase
Description
General description
Three ports. Greaseless vacuum line fabricated with Ace-Threds and PTFE stopcocks. Stopcocks are parallel to main body, permitting operation from front or back. At top of main body is adapter for connection to thermocouple vacuum gauge.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?