• USA Home
  • Z160059 - Manipulation of Air-Sensitive Compounds, 2nd Ed.
EMAIL THIS PAGE TO A FRIEND
Z160059 Aldrich
Manipulation of Air-Sensitive Compounds, 2nd Ed.
  •  ISBN-10 047186773X
  •  ISBN-13 978-047186773-9
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   D. Shriver and M. Drezdzon, John Wiley & Sons, 1986, 336 pp., hard cover
mfr. no.   Wiley
Description
General description
Revised to reflect the continuing and growing importance of research and development within this field, this second edition offers state-of-the-art methods used in handling air-sensitive compounds, including gases. Part One covers inert atmosphere techniques, while Part Two treats vacuum line techniques. Appendixes provide safety data, information on materials used to construct apparatus, and a table of vapor pressures of common volatile substances.
Table of Contents
INERT ATMOSPHERE TECHINQUES.
Bench-Top Inert-Atmosphere Techniques.
Inert-Atmosphere Glove Boxes.
Inert Gases and Their Purification.
Purification of Solvents and Reagents.
Vacuum Line Design and Operation.
Pumps for Rough and High Vacuum.
Pressure and Flow Measurement and Leak Detection.
Joints, Stopcocks, and Valves.
Specialized Vacuum-Line Equipment and Operations.
Metal Systems.
Metal Systems.
Appendixes.
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?