EMAIL THIS PAGE TO A FRIEND
Z160253 Aldrich
Universal timer
60 hr
Purchase
Properties
Related Categories Labware, Timers More...
feature   subdivision minutes
measuring range   60 hr
Description
General description
Features include:
±0.015% accuracy at maximum dial setting; 6 1/2 in. diam. dial with large numerals for elapsed time and an inner circle of smaller numerals for use as a stopclock. Built-in buzzer with variable tone control; PC/PET case that measures 7 1/2 × 7 1/2 × 2 1/2 in. and has rubber base pads, wall mounting keyholes, and recessed hand grip for portability. Two separate 3-wire outlet receptacles for automatic switching of external appliances are provided (one is controlled by a switch and always breaks circuit at "zero").
Specifications:
Input voltage of 100 to 130 V AC, 50 or 60Hz; power rating of AC outlets at 120 V AC: max. lamp load of 600 V; max. resistive load of 1,200 W; max. motor load of Ghp. UL and CSA listed.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?