• USA Home
  • Z160377 - Thermometer for MEL-TEMP® apparatus
EMAIL THIS PAGE TO A FRIEND
Z160377 Aldrich
Thermometer for MEL-TEMP® apparatus
Fluke®
digital thermometer
Purchase
Properties
Related Categories Fluke, Labware, Manufacturer Browser, Melting Point Apparatus, Temperature,
mfr. no.   Fluke
H × W × L   1 in. × 3 in. × 6 in.
Description
General description
• Use with Type-K or J thermocouple probes
• Select readout in °C or °F and convert anytime during operation
• Push-button controls for all functions
• HOLD button freezes display; push again to release
• 0.1 °C (0.2 °F) resolution over entire measurement range (4-digits)
• Includes general-purpose bead thermocouple (not suitable for liquid immersion) and 9 V battery
Legal Information
Fluke is a registered trademark of John Fluke Manufacturing Co., Inc.
MEL-TEMP is a registered trademark of Barnstead/Thermolyne Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
92451 Timestrip Plus 25 °C
08168 Timestrip Plus 8 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?