• USA Home
  • Z160393 - Fluke® Type-K bead probe
EMAIL THIS PAGE TO A FRIEND
Z160393 Aldrich
Fluke® Type-K bead probe
parameter -40-260 °C temperature (-40-500 °F)
Purchase
Properties
Related Categories Fluke, Labware, Manufacturer Browser, Temperature, Thermocouples More...
mfr. no.   Fluke
parameter   -40-260 °C temperature (-40-500 °F)
Description
General description
General purpose applications, PTFE tape insulation. Not suitable for liquid immersion.
Legal Information
Fluke is a registered trademark of John Fluke Manufacturing Co., Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
92451 Timestrip Plus 25 °C
08168 Timestrip Plus 8 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?