• USA Home
  • Z160407 - Fluke® Type-K immersion probe
EMAIL THIS PAGE TO A FRIEND
Z160407 Aldrich
Fluke® Type-K immersion probe
parameter -40-1800 °F temperature
Purchase
Properties
Related Categories Fluke, Labware, Manufacturer Browser, Temperature, Thermocouples More...
mfr. no.   Fluke
parameter   -40-1800 °F temperature
Description
General description
For liquids and gels. Protected by Inconel sheath, can be used in most general temperature measurement applications.
Legal Information
Fluke is a registered trademark of John Fluke Manufacturing Co., Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
92451 Timestrip Plus 25 °C
08168 Timestrip Plus 8 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?