• USA Home
  • Z162116 - Water-jet aspirator pump
EMAIL THIS PAGE TO A FRIEND
Z162116 Aldrich
Water-jet aspirator pump
Without pressure gauge
Purchase
Properties
Related Categories Labware, Pump Aspirators, Pumps More...
material   polypropylene
mfr. no.   Azlon
Description
General description
Side-arm tapers from 7 to 12 mm with an integral check valve. Operates effectively (half volume) at a low flow rate of 7 L/min. Max. vacuum is 25 mm Hg (abs) at a flow rate of 17 L/min and a water pressure of 2.8 kg/cm2.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z150762 Water aspirator pump, Short swirling
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?