• USA Home
  • Z162914 - Whirl-pak® sampling bag
EMAIL THIS PAGE TO A FRIEND
Z162914 Aldrich
Whirl-pak® sampling bag
Sampling, capacity 540 mL
Purchase
Properties
Related Categories Bags, Labware More...
label   A
W × L   4.5 in. (11.5 cm) × 9 in. (23 cm)
capacity   540 mL
thickness   2.5 mil
Description
General description
Heavy, transparent, non-toxic, sterilized HDPE. Double pull-tab for easy opening and double-wire tab for closing. Leak proof and airtight. Easily labeled with waterproof pen.
Legal Information
Whirl-pak is a registered trademark of The Aristotle Corporation
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?