EMAIL THIS PAGE TO A FRIEND
Z163368 Aldrich
Desiccant paper
Purchase
Properties
Related Categories Desiccants, Desiccators and Desiccants, Labware More...
Description
General description
Comprised of cellulose fiber and silica gel; 67% of weight is silica gel. Easily reactivated (1 to 5 min at 300 °F, 5 to 10 min at 300 °F). 12 × 12 in. sheets, 54 mil thick. Dry weight of sheet is ~73 g. In packages of 5 sheets; sealed in barrier pouch.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?