• USA Home
  • Z163996 - Aldrich® ‘N′-Tube
EMAIL THIS PAGE TO A FRIEND
Z163996 Aldrich
Aldrich® ‘N′-Tube
female Joint: ST/NS 24/40
Purchase
Properties
Related Categories ‘N′-Tube, Aldrich, Aldrich Distillation Equipment, Aldrich Distillation Traps, Dean-Stark Glass Distillation Traps,
Joint   female Joint: ST/NS24/40
  male Joint ST/NS24/40
availability   not available in EU
capacity   10 mL
Description
General description
For small scale distillation.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?