• USA Home
  • Z164399 - Twistit four-in-one rubber stoppers
EMAIL THIS PAGE TO A FRIEND
Z164399 Aldrich
Twistit four-in-one rubber stoppers
size 9
Synonym: rubber plug, rubber plugs, rubber stopper
Purchase
Properties
Related Categories Labware, Rubber Stoppers, Stoppers More...
size   9
Description
General description
Twist-off seals provide multiple uses of one stopper. Made of soft, white, natural rubber having high resistance to deformation after autoclaving. Has lead countersunk into holes and is vacuum tested at 30 mm Hg.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?