• USA Home
  • Z165522 - Double-needle liquid-transfer/gas-purge unit
EMAIL THIS PAGE TO A FRIEND
Z165522 Aldrich
Double-needle liquid-transfer/gas-purge unit
L 6 in., size 20 gauge
Purchase
Description
General description
304 SS needles with noncoring points and female Luer hub. Two needles soldered together are more rigid and allow fewer vapors to be released through septum puncture during transfer or purge. Second needle is always 1in. long. Stated length is "x" in drawing.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?