• USA Home
  • Z166863 - TLC sample recovery tube
EMAIL THIS PAGE TO A FRIEND
Z166863 Aldrich
TLC sample recovery tube
bulb capacity 1-2 mL
Purchase
Properties
Related Categories Analytical/Chromatography, LPLC/TLC/Paper Chromatography, Pipettes, TLC Accessories, Thin Layer Chromatography (TLC) More...
bulb capacity   1-2 mL
pore size   10-15 μm porosity
Description
General description
For recovery of samples directly from developed TLC plates by suction. Recovery is by elution with appropriate solvent while absorbent is retained on fritted disc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?