• USA Home
  • Z167940 - Glass water aspirator pump
EMAIL THIS PAGE TO A FRIEND
Z167940 Aldrich
Glass water aspirator pump
100 mmHg (abs.) Max. vacuum
Purchase
Properties
Related Categories Kimble Kontes, Kimble Kontes Pumps, Labware, Manufacturer Browser, Pump Aspirators,
feature   100 mmHg (abs.) Max. vacuum
mfr. no.   Kimble Kontes
Description
General description
Suitable for use as a backing pump in distillation or filtration.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?