• USA Home
  • Z168440 - Thermolyne® Cimarec®-top hotplate
EMAIL THIS PAGE TO A FRIEND
Z168440 Aldrich
Thermolyne® Cimarec®-top hotplate
AC input 120 V, plate size 7 in. × 7 in.
Purchase
Properties
Related Categories Barnstead Thermolyne, Barnstead Thermolyne Heating Equipment, Barnstead Thermolyne Hot Plates, Heating, Hotplates,
sterility   non-sterile
mfr. no.   Barnstead International HP46825
availability   not available in EU
plate size   7 in. × 7 in.
output   1100 watts
matrix active group   Non-treated surface treatment
Description
General description
Solid ceramic top has high impact strength, acid and alkali resistance, and over-hanging design prevents spills from entering unit. Adjustable temp. range to max. 538 °C (1,000 °F) within 8 min. Cast aluminum body. All units CSA listed.
Legal Information
Cimarec is a registered trademark of Barnstead/Thermolyne Corp.
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?