• USA Home
  • Z168491 - Thermolyne® Cimarec®-top magnetic stirrer
EMAIL THIS PAGE TO A FRIEND
Z168491 Aldrich
Thermolyne® Cimarec®-top magnetic stirrer
top plate size 4 in. × 4 in., AC/DC input 240 V AC, 0.03 A, output80 W
Purchase
Properties
Related Categories Labware, Magnetic Stirrers, Stirrers More...
AC/DC input   50 - 60 Hz
feature   CE compliant
mfr. no.   Barnstead International S46410-33
speed   100-1000 rpm
top plate size   4 in. × 4 in.
output   80 W
Description
General description
Has pure-ceramic solid top.
Legal Information
Cimarec is a registered trademark of Barnstead/Thermolyne Corp.
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?