• USA Home
  • Z168505 - Thermolyne® Cimarec®-top magnetic stirrer
EMAIL THIS PAGE TO A FRIEND
Z168505 Aldrich
Thermolyne® Cimarec®-top magnetic stirrer
top plate size 7 in. × 7 in., AC input 120 V, 0.15 A, output18 W
Purchase
Properties
Related Categories Barnstead Thermolyne, Barnstead Thermolyne Stirrers, Labware, Magnetic Stirrers, Manufacturer Browser,
AC input   50 - 60 Hz
feature   CE compliant
mfr. no.   Barnstead International S46725
speed   100-1000 rpm
top plate size   7 in. × 7 in.
output   18 W
Description
General description
Has pure-ceramic solid top.
Legal Information
Cimarec is a registered trademark of Barnstead/Thermolyne Corp.
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
CLS6795210 Corning® plate stirrers, 4 x 5 inch top PC-210 stirrer, pyroceram glass-ceramic top, 120V/60Hz, 1/cs
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?