• USA Home
  • Z168556 - Thermolyne® Cimarec®-top stirring hotplate
EMAIL THIS PAGE TO A FRIEND
Z168556 Aldrich
Thermolyne® Cimarec®-top stirring hotplate
AC input 120 V, plate size 4 in. × 4 in.
Purchase
Properties
Related Categories Barnstead Thermolyne, Barnstead Thermolyne Heating Equipment, Barnstead Thermolyne Hot Plates, Heating, Hotplate Stirrers,
sterility   non-sterile
feature   CSA approved
mfr. no.   Barnstead International SP46615
availability   not available in EU
plate size   4 in. × 4 in.
output   388 watts
matrix active group   Non-treated surface treatment
Description
General description
Has all features of the Cimarec-top hot plate plus has excellent low-speed stirring control and speed range of 60 to 1,000 rpm. All units are CSA listed.
Legal Information
Cimarec is a registered trademark of Barnstead/Thermolyne Corp.
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?