• USA Home
  • Z169374 - Spectroline® CM UV-viewing cabinet
EMAIL THIS PAGE TO A FRIEND
Z169374 Aldrich
Spectroline® CM UV-viewing cabinet
Cabinet CM-10
Purchase
Description
General description
Portable, lightweight aluminum minicabinets with 4 and 6 W removable lamps that may be used for hand-held applications. Lamps provide long- and shortwave irradiation for viewing TLC plates, and for UV fluorescence or absorption demonstrations. Cabinets have a flexible, contoured viewer for built-in UV-absorbing window, easy-access felt curtains, and removable bottoms for placement over transilluminators for greater intensity and contrast. CE compliant, UL listed, and meets OSHA requirements.
CM-24: cabinet with removable, combination UV light source (365nm/254nm); 4-watt tubes
CM-26: cabinet with removable, combination UV light source (365nm/254nm); 6-watt tubes
Without light source. May be used with 4 and 6 watt Spectroline UV lamps.
Legal Information
Spectroline is a registered trademark of Spectronics, Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?