• USA Home
  • Z169943 - Scienceware® glove box
EMAIL THIS PAGE TO A FRIEND
Z169943 Aldrich
Scienceware® glove box
Purchase
Properties
Related Categories Bel-Art, Bel-Art Glove Bags & Boxes, Glove Boxes, Labware, Manufacturer Browser,
mfr. no.   Bel-Art
H × L × D   26 in. × 36 in. × 20 in.
thickness   5/16 in.
Description
General description
Clear PMMA glove box resists most inorganic solutions, oils, low concentrations of alcohols, paraffinic and olefinic hydrocarbons, amines and alkyl monohalides, and esters containing more than 10 carbon atoms. Connectable to other glove boxes. Two side openings with 7/8 in. flange, 12 in. O.D. and 113/4 in. I.D. with PVC gaskets on outside, and removable covers. Glove ports (8 in. O.D. and 73/4 in. I.D. with 1 in. external flange) accept standard dry-box gloves (not included) or barehand entry (iris port), Z170119.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
accessory
Product #
Description
Add to Cart
Z169951 Scienceware® glove box air lock, pkg of 1 ea
related product
Product #
Description
Add to Cart
Z169994 Scienceware® glove box hand-port cover, port diam. 20.3 cm, Z169943
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?