Z169951 Aldrich
Scienceware® glove box air lock
pkg of 1 ea
DOWNLOAD MSDS (PDF)
Synonym: humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
Properties
Related Categories Bel-Art, Bel-Art Glove Bags & Boxes, Glove Boxes, Labware, Manufacturer Browser,
label   A
packaging   pkg of 1 ea
Description
General description
The Scienceware glove box air lock is supplied with connecting sleeve with 12 in. diam. interior cover to separate air lock from glove box. Side-entry cover of glove box becomes outside door of air lock. The air lock is used with the Scienceware glove box, Z169943.
Legal Information
Scienceware is a registered trademark of Bel-Art Products
Price and Availability
Customers Also Viewed
Plas-Labs basic glove box
AC input 240 V, Euro plug, Plas-Labs, 818-GB/EXP
port diam. 20.3 cm, Z169943
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?