• USA Home
  • Z169986 - Scienceware® glove box coupling sleeve
EMAIL THIS PAGE TO A FRIEND
Z169986 Aldrich
Scienceware® glove box coupling sleeve
Purchase
Properties
Related Categories Bel-Art, Bel-Art Glove Bags & Boxes, Glove Boxes, Labware, Manufacturer Browser,
label   B
mfr. no.   Bel-Art
Description
General description
For connection of two or more glove boxes in series, or for replacement of air lock.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?