• USA Home
  • Z170283 - Lightnin® 316 stainless steel impeller
EMAIL THIS PAGE TO A FRIEND
Z170283 Aldrich
Lightnin® 316 stainless steel impeller
3 Blade turbine, Model A-310, diam. 4 1/2 in. , overall
Purchase
Properties
Related Categories Labware, Lightnin, Manufacturer Browser, Overhead - SS Stirrers, Stirrers More...
mfr. no.   Lightnin 809412
  , Model A-310
bore size   5/16 in.
diam.   4 1/2 in. , overall
Description
General description
High efficiency design develops 50% more mixing action and true axial flow.
Legal Information
Lightnin is a registered trademark of Lightnin, a unit of General Signal
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?