• USA Home
  • Z170399 - Lightnin® 316 stainless steel impeller
EMAIL THIS PAGE TO A FRIEND
Z170399 Aldrich
Lightnin® 316 stainless steel impeller
Folding propeller, diam. 2.0 in. , overall
Purchase
Properties
Related Categories Labware, Lightnin, Manufacturer Browser, Overhead - SS Stirrers, Stirrers More...
mfr. no.   Lightnin 800578
  , Model A-100
bore size   5/16 in.
diam.   2.0 in. , overall
Description
General description
For vessels with axial narrow openings, folded diam. is 11/16 in., opens for full mixing action.
Legal Information
Lightnin is a registered trademark of Lightnin, a unit of General Signal
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?