• USA Home
  • Z171557 - Series A gas syringe
EMAIL THIS PAGE TO A FRIEND
Z171557 Aldrich
Series A gas syringe
capacity 5 mL
Purchase
Properties
Related Categories Labware, Manufacturer Browser, Series A gas syringe, VICI
capacity   5 mL
Description
General description
Same as series A-2 gas syringes above, but with turn valve instead of push-button valve.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?