• USA Home
  • Z173312 - Aldrich® Schlenk-type flasks and tubes
EMAIL THIS PAGE TO A FRIEND
Z173312 Aldrich
Aldrich® Schlenk-type flasks and tubes
100ml, S/T14/20M joint
Purchase
Properties
Related Categories Air-Sensitive Chemistry, Air-Sensitive Glassware, Aldrich Air Sensitive, Aldrich Air Sensitive Schlenk-Type, Aldrich Air Sensitive Schlenk-Type Flasks,
material   borosilicate glass
sterility   non-sterile
feature   closure type Screw cap
packaging   pack of 1
mfr. no.   Sigma-Aldrich
capacity   100 mL
Description
General description
Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?