• USA Home
  • Z173339 - Aldrich® Schlenk-type adapter
EMAIL THIS PAGE TO A FRIEND
Z173339 Aldrich
Aldrich® Schlenk-type adapter
Connecting, female Joints: threaded 14/20 (2), male Joint: threaded 14/20
Purchase
Properties
Related Categories Air-Sensitive Chemistry, Aldrich Air Sensitive, Aldrich Air Sensitive Schlenk-Type, Aldrich Air Sensitive Schlenk-Type Adapters and Caps, Labware More...
Joint   female Joints: threaded14/20 (2)
  male Joint: threaded14/20
Description
General description
Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?