• USA Home
  • Z173355 - Low temperature filter funnel
EMAIL THIS PAGE TO A FRIEND
Z173355 Aldrich
Low temperature filter funnel
capacity 75 mL, male Joint: ST/NS 14/20, female Joint: ST/NS 14/20
Purchase
Properties
Related Categories Air-Sensitive Chemistry, Air-Sensitive Glassware, Aldrich Air Sensitive, Aldrich Air Sensitive Schlenk-Type, Aldrich Air Sensitive Schlenk-Type Funnels,
label   Z
Joint   female Joint: ST/NS14/20
  male Joint: ST/NS14/20
capacity   75 mL
pore size   25-50 μm porosity
Description
General description
Lower outlet is bent to allow for immersion of entire unit into a Dewar bath.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?