• USA Home
  • Z173460 - Aldrich® Schlenk funnels and filters
EMAIL THIS PAGE TO A FRIEND
Z173460 Aldrich
Aldrich® Schlenk funnels and filters
capacity 75 mL, Filter tube with PTFE stopcock, male Joints: ST/NS 14/20 (2), female Joints: ST/NS 14/20 (2)
Purchase
Properties
Related Categories Air-Sensitive Chemistry, Air-Sensitive Glassware, Aldrich Air Sensitive, Aldrich Air Sensitive Schlenk-Type, Aldrich Air Sensitive Schlenk-Type Filters,
Joint   female Joints: ST/NS14/20 (2)
  male Joints: ST/NS14/20 (2)
capacity   75 mL
Description
General description
Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?