• USA Home
  • Z173576 - Bonnet-vent tube connector
EMAIL THIS PAGE TO A FRIEND
Z173576 Aldrich
Bonnet-vent tube connector
for tubing o.d., 1/4 in.
Purchase
Properties
Related Categories Adapters, Aldrich, Aldrich Adapters (Gas), Aldrich Gas Equipment, Gas Equipment Adapters,
material   316 stainless steel
feature   for tubing o.d. 1/4 in.
fitting   1/8 in. male NPT
Description
General description
Use to attach vent tube to gas regulators with bonnet-vent fittings.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?