• USA Home
  • Z173614 - Sample cylinder stainless steel
EMAIL THIS PAGE TO A FRIEND
Z173614 Aldrich
Sample cylinder stainless steel
capacity 300 mL
Purchase
Description
General description
With two 1/4 in. NPTF end valve threads. Can be used to store or transport liquids and gases at pressures up to 1,800 psig at room temperature; also, as pressure purge accumulator, reaction vessel, gas sampling for chromatography, lecture bottle for lab use, and for condensate sampling and transporting in fossil fuel and nuclear power plants.
Hot-spun (not welded) from seamless, 304 grade SS tubing for maximum integrity. Corrosion resistant. Raw material is heat code traceable.
These cylinders comply with DOT-3E 1800 (US), which states it can contain flammable liquids or gases during transport. Not exceeding the pressure rating of the vessel. These cylinders also comply with TPED overseas (Europe).
Order two SS valves, Z146064, Z146056, or Z168637 separately. DOT 3E-1800.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?