• USA Home
  • Z174319 - Single-bulb ball-tube
EMAIL THIS PAGE TO A FRIEND
Z174319 Aldrich
Single-bulb ball-tube
capacity 4,000 mL, Joint: ST/NS 24/40, plain
Purchase
Description
General description
Arranged horizontally outside of Kugelrohr oven, the ball-tubes cool and collect distillate with minimal loss of material.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?