• USA Home
  • Z174378 - Disposable gas and liquid in-line filter
EMAIL THIS PAGE TO A FRIEND
Z174378 Aldrich
Disposable gas and liquid in-line filter
liquid efficiency 8 μm, 98% retention
Purchase
Properties
packaging   pkg of 10
pore size   0.1 μm gas efficiency, 98% retention
  8 μm liquid efficiency, 98% retention
Description
General description
For gas and liquid applications. Leak-proof in-line filter suitable for use with toxic and radioactive materials. Consists of microfiber filter tube bonded into sealed clear nylon holder with a 125psig pressure rating and 1/4 in. O.D. ports for pressure-tight connections using standard compression fittings. For low-pressure applications, slip-on tubing or plastic barbs may be used.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?