• USA Home
  • Z175692 - Aldrich® Oldershaw distilling column
EMAIL THIS PAGE TO A FRIEND
Z175692 Aldrich
Aldrich® Oldershaw distilling column
no. of plates, 5, I.D. × O.D. 28 mm × 64 mm, Joints: ST/NS 29/42 (2)
Purchase
Description
General description
Vacuum-jacketed, strip-silvered to provide two windows 180° apart for viewing. Consists of a series of sealed-in, perforated plates, each of which has a drain tube to direct condensed distillate to next lower plate and a weir to maintain uniform liquid level.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?