• USA Home
  • Z175765 - Strip-silvered Hempel columns
EMAIL THIS PAGE TO A FRIEND
Z175765 Aldrich
Strip-silvered Hempel columns
L 385 mm
Purchase
Properties
Related Categories Aldrich, Aldrich Distillation Columns, Aldrich Distillation Equipment, Aldrich Hempel Columns, Glass Distillation Columns,
Joint   Joint ST/NS24/40
I.D.   19 mm
L   385 mm
O.D.   42 mm
Description
General description
Vacuum-jacketed and strip-silvered to provide two windows 180° apart for viewing (except the 160 mm column which is completely silvered).
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?