• USA Home
  • Z175811 - Aldrich® vacuum-jacketed Buchner funnel
EMAIL THIS PAGE TO A FRIEND
Z175811 Aldrich
Aldrich® vacuum-jacketed Buchner funnel
Joint: ST/NS 24/40
Purchase
Properties
Related Categories Glass Buchner Funnels, Glass Funnels, Glassware, Labware
Joint   Joint: ST/NS24/40
Description
General description
Silvered, 60 mL capacity, 180 mm H with 40 mm diam. frit, 25 to 50μm porosity. Drip-tip joint and vacuum take-off.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?